missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Fumaric acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002411
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002411 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002411 Supplier Merck Emd Millipore Supplier No. PHL80141100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 110-17-8
Product Line phyproof
Ignition Point 1364°F
Vapor Pressure 1.7 mmHg (165°C)
Flash Point 272°C to 274°C - open cup
Linear Formula HOOCCH=CHCOOH
MDL Number MFCD00002700
Melting Point 298°C to 300°C (sublimation)
Type Reference Substance
Quantity 100 mg
Formula Weight 116.07
Percent Purity ≥98% (HPLC)
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG