accessibility menu, dialog, popup

UserName

CH3CH3H3CSSCH3CH3H3C

CAS RN 110-06-5

IUPAC Name: 2-(tert-butyldisulfanyl)-2-methylpropane
Synonyms: di-tert-butyldisulfide
Molecular Weight (g/mol): 178.35
Molecular Formula: C8H18S2
InChi Key: BKCNDTDWDGQHSD-UHFFFAOYSA-N
SMILES: CC(C)(C)SSC(C)(C)C
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)