CAS RN 109945-55-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lewatit™ MonoPlus TP 214 Ion Exchange Resin, MilliporeSigma™ Supelco™
Lewatit™ MonoPlus TP 214 is a type of chelating ion-exchange resin.