accessibility menu, dialog, popup

UserName

H3CNCH3SOOHNOHO

CAS RN 1091-85-6

IUPAC Name: 2-[5-(dimethylamino)naphthalene-1-sulfonamido]acetic acid
Synonyms: [5-(dimethylamino)naphthalene-1-sulfonamido]acetic acid
Molecular Weight (g/mol): 308.35
Molecular Formula: C14H16N2O4S
InChi Key: QHFXORCWAQTTGH-UHFFFAOYSA-N
SMILES: CN(C)C1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NCC(O)=O
Percent Purity 
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results