accessibility menu, dialog, popup

UserName

CAS RN 109-67-1

IUPAC Name: pent-1-ene
Synonyms: pentene
Molecular Weight (g/mol): 70.14
Molecular Formula: C5H10
InChi Key: YWAKXRMUMFPDSH-UHFFFAOYSA-N
SMILES: CCCC=C
Percent Purity 
  • (2)
  • (1)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (2)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (7)