missing translation for 'onlineSavingsMsg'
Learn More

1-Pentene, 97%

Catalog No. AC268661000
Change view
Click to view available options
Quantity:
25 mL
100 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268661000 100 mL Glass bottle
AC268660250 25 mL Glass bottle
AC268660025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC268661000 Supplier Thermo Scientific Chemicals Supplier No. 268661000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-67-1 | C5H10 | 70.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-67-1
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00003567
InChI Key YWAKXRMUMFPDSH-UHFFFAOYSA-N
Synonym 1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
PubChem CID 8004
IUPAC Name pent-1-ene
SMILES CCCC=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -165.0°C
Color Colorless
Density 0.6400g/mL
Boiling Point 30.0°C
Flash Point −28°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3CH2CH2CH=CH2
Refractive Index 1.3704 to 1.3724
Quantity 100 mL
Beilstein 01,21
Merck Index 15,7234
Specific Gravity 0.64
Solubility Information Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether and beznene, soluble in most organic solvents
Viscosity 0.202 mPa.s (20°C)
Formula Weight 70.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Pentene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Ta
EINECSNumber 203-694-5

RUO – Research Use Only