accessibility menu, dialog, popup

UserName

CAS RN 1086-00-6

Cl

CAS RN 1086-00-6

IUPAC Name: 1-(chloromethyl)pyrene
Synonyms: 1-(chloromethyl)pyrene
Molecular Weight (g/mol): 250.73
Molecular Formula: C17H11Cl
InChi Key: MVNXSXOJYGSNQZ-UHFFFAOYSA-N
SMILES: ClCC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results