missing translation for 'onlineSavingsMsg'
Learn More

1-Chloromethylpyrene 97.0+%, TCI America™

Catalog No. C16371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16371G Supplier TCI America Supplier No. C16371G
Only null left

Chemical Identifiers

CAS 1086-00-6
Molecular Formula C17H11Cl
Molecular Weight (g/mol) 250.725
MDL Number MFCD00962113
InChI Key MVNXSXOJYGSNQZ-UHFFFAOYSA-N
PubChem CID 97581
IUPAC Name 1-(chloromethyl)pyrene
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)CCl

Specifications

Quantity 1 g
Formula Weight 250.73
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloromethylpyrene
RTECSNumber UR2450700
TSCA No
Recommended Storage Freezer