accessibility menu, dialog, popup

UserName

ONON

CAS RN 1083-48-3

IUPAC Name: 4-[(4-nitrophenyl)methyl]pyridine
Synonyms: 4-[(4-nitrophenyl)methyl]pyridine
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: MNHKUCBXXMFQDM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CC2=CC=NC=C2)C=C1
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)