accessibility menu, dialog, popup

UserName

CAS RN 108238-09-1

IUPAC Name: (2-phenoxyphenyl)boronic acid
Synonyms: 2-phenoxyphenylboronic acid
Molecular Weight (g/mol): 214.03
Molecular Formula: C12H11BO3
InChi Key: AVOWPOFIQZSVGV-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results