accessibility menu, dialog, popup

UserName

CAS RN 1081-71-6

IUPAC Name: 3-(4-hydroxy-3-methoxyphenyl)-2-oxopropanoic acid
Synonyms: 3-(4-hydroxy-3-methoxyphenyl)-2-oxopropanoic acid
Molecular Weight (g/mol): 210.19
Molecular Formula: C10H10O5
InChi Key: YGQHQTMRZPHIBB-UHFFFAOYSA-N
SMILES: COC1=CC(CC(=O)C(O)=O)=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results