accessibility menu, dialog, popup

UserName

CH3NOHNCH3

CAS RN 108-79-2

IUPAC Name: 4,6-dimethyl-1,2-dihydropyrimidin-2-one
Synonyms: 4,6-dimethyl-1H-pyrimidin-2-one
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: WHEQVHAIRSPYDK-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(=O)N1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results