accessibility menu, dialog, popup

UserName

CAS RN 108-68-9

IUPAC Name: 3,5-dimethylphenol
Synonyms: 3,5-dimethylphenol xylenol 200 xylenol (substance) ccris 724 1,3,5-xylenol sym-m-xylenol 1,5-dimethyl-3-hyperoxybenzene
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: TUAMRELNJMMDMT-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(C)=C1
Boiling Point 
  • (2)
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (6)
  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (4)