accessibility menu, dialog, popup

UserName

CAS RN 108-67-8

IUPAC Name: 1,3,5-trimethylbenzene
Synonyms: mesitylene trimethylbenzol fleet-X TMB s-trimethylbenzene sym-trimethylbenzene hsdb 92
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: AUHZEENZYGFFBQ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=CC(C)=C1
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Grade 
  • (1)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (2)
  • (8)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (7)