accessibility menu, dialog, popup

UserName

CAS RN 108-42-9

IUPAC Name: 3-chloroaniline
Synonyms: 3-chloroaniline
Molecular Weight (g/mol): 127.57
Molecular Formula: C6H6ClN
InChi Key: PNPCRKVUWYDDST-UHFFFAOYSA-N
SMILES: NC1=CC=CC(Cl)=C1
Percent Purity 
  • (2)
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)