missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroaniline, 99%

Catalog No. AC108585000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108585000 500 mL Glass bottle
AC108581000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108585000 Supplier Thermo Scientific Chemicals Supplier No. 108585000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-42-9 | C6H6ClN | 127.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-42-9
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00007765
InChI Key PNPCRKVUWYDDST-UHFFFAOYSA-N
Synonym m-chloroaniline, 3-chlorobenzenamine, benzenamine, 3-chloro, m-chloraniline, 3-chlorophenylamine, orange gc base, m-chlorophenylamine, m-aminochlorobenzene, m-chloroaminobenzene, aniline, m-chloro
PubChem CID 7932
IUPAC Name 3-chloroaniline
SMILES NC1=CC=CC(Cl)=C1

Specifications

Melting Point -11.0°C
Color Yellow to Brown
CAS Min % 98.5
CAS Max % 100.0
Density 1.2000g/mL
Quantity 500 mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5927 to 1.5947
Specific Gravity 1.2
Assay Percent Range 98.5% min. (GC)
Linear Formula ClC6H4NH2
Beilstein 12, 602
Merck Index 15, 2118
Boiling Point 230.0°C
Solubility Information Solubility in water: 6.8g/L (20°C). Other solubilities: soluble in most common organic solvents
Formula Weight 127.57
Percent Purity 99%
Chemical Name or Material 3-Chloroaniline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
EINECSNumber 203-581-0
RTECSNumber BX0350000
TSCA TSCA
Recommended Storage The product turns dark during prolonged storage

RUO – Research Use Only