missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloroaniline, 99%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS | 108-42-9 |
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Density | 1.2000g/mL |
| Boiling Point | 230.0°C |
| Flash Point | 118°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Molecular Formula | C6H6ClN |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic in contact with skin. Toxic if inhaled. Toxic if swallowed. May cause damage to organs through prolonged or repeated exposure if inhaled. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove |
| EINECSNumber | 203-581-0 |
| RTECSNumber | BX0350000 |
| TSCA | TSCA |
| Recommended Storage | The product turns dark during prolonged storage |
RUO – Research Use Only