missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroaniline 99.0+%, TCI America™

Catalog No. C0110500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0110500ML 500 mL
C011025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0110500ML Supplier TCI America Supplier No. C0110500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-42-9
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00007765
InChI Key PNPCRKVUWYDDST-UHFFFAOYSA-N
Synonym m-chloroaniline, 3-chlorobenzenamine, benzenamine, 3-chloro, m-chloraniline, 3-chlorophenylamine, orange gc base, m-chlorophenylamine, m-aminochlorobenzene, m-chloroaminobenzene, aniline, m-chloro
PubChem CID 7932
IUPAC Name 3-chloroaniline
SMILES NC1=CC=CC(Cl)=C1

Specifications

Melting Point -10°C
Color Yellow
Quantity 500 mL
Physical Form Clear Liquid at 20°C
UN Number 2019
Boiling Point 230°C
Formula Weight 127.57
Percent Purity ≥99.0% (GC)
Chemical Name or Material 3-Chloroaniline
EINECSNumber (3)-0194
RTECSNumber BX0350000
TSCA Yes