accessibility menu, dialog, popup

UserName

CAS RN 1078-71-3

IUPAC Name: heptylbenzene
Synonyms: 1-phenylheptane
Molecular Weight (g/mol): 176.3
Molecular Formula: C13H20
InChi Key: LBNXAWYDQUGHGX-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)