CAS RN 1073234-31-7
IUPAC Name:
[2-(benzyloxy)phenyl]methyl acetate
Synonyms:
[2-(benzyloxy)phenyl]methyl acetate
Molecular Weight (g/mol):
256.3
Molecular Formula:
C16H16O3
InChi Key:
AEQBBYFCUOIBIW-UHFFFAOYSA-N
SMILES:
CC(=O)OCC1=CC=CC=C1OCC1=CC=CC=C1
1
–
1
of
1
results
1
2-Benzyloxybenzyl Acetate 98.0+%, TCI America™
CAS: 1073234-31-7 Molecular Formula: C16H16O3 Molecular Weight (g/mol): 256.301 InChI Key: AEQBBYFCUOIBIW-UHFFFAOYSA-N Synonym: Acetic Acid 2-Benzyloxybenzyl Ester PubChem CID: 44629830 IUPAC Name: (2-phenylmethoxyphenyl)methyl acetate SMILES: CC(=O)OCC1=CC=CC=C1OCC2=CC=CC=C2