accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNOCH3

CAS RN 1072945-01-7

IUPAC Name: 2-ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 249.12
Molecular Formula: C13H20BNO3
InChi Key: GXUHWEZZHFBDFA-UHFFFAOYSA-N
SMILES: CCOC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results