accessibility menu, dialog, popup

UserName

H2NOPOHOHO

CAS RN 1071-23-4

IUPAC Name: (2-aminoethoxy)phosphonic acid
Synonyms: O-phosphoethanolamine colaminphosphoric acid phosphorylethanolamine magnesium (1:1) salt EAP OPE PEA phosphorylethanolamine phosphoryethanolamine ethanolamine acid phosphate ethanolamine O-phosphate
Molecular Weight (g/mol): 141.06
Molecular Formula: C2H8NO4P
InChi Key: SUHOOTKUPISOBE-UHFFFAOYSA-N
SMILES: NCCOP(O)(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)