accessibility menu, dialog, popup

UserName

CAS RN 107017-73-2

H3CCH3H3COOHNHO

CAS RN 107017-73-2

IUPAC Name: tert-butyl N-[1-(hydroxymethyl)cyclopropyl]carbamate
Synonyms: tert-butyl N-[1-(hydroxymethyl)cyclopropyl]carbamate
Molecular Weight (g/mol): 187.24
Molecular Formula: C9H17NO3
InChi Key: HFMAZNJKNNRONT-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1(CO)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)