accessibility menu, dialog, popup

UserName

CAS RN 107-81-3

IUPAC Name: 2-bromopentane
Synonyms: 2-bromopentane
Molecular Weight (g/mol): 151.05
Molecular Formula: C5H11Br
InChi Key: LGAJYTCRJPCZRJ-UHFFFAOYNA-N
SMILES: CCCC(C)Br
Boiling Point 
  • (1)
  • (2)
Grade 
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)