accessibility menu, dialog, popup

UserName

ClH2C

CAS RN 107-05-1

IUPAC Name: 3-chloroprop-1-ene
Synonyms: allyl chloride
Molecular Weight (g/mol): 76.52
Molecular Formula: C3H5Cl
InChi Key: OSDWBNJEKMUWAV-UHFFFAOYSA-N
SMILES: ClCC=C
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)