accessibility menu, dialog, popup

UserName

CH3OPOH3COOH3C

CAS RN 1067-71-6

IUPAC Name: diethyl (2-oxopropyl)phosphonate
Synonyms: diethyl 2-oxopropylphosphonate
Molecular Weight (g/mol): 194.17
Molecular Formula: C7H15O4P
InChi Key: RSAFKRSMGOSHRK-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC(C)=O)OCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)