accessibility menu, dialog, popup

UserName

CAS RN 106562-32-7

IUPAC Name: 2-(7-amino-4-methyl-2-oxo-2H-chromen-3-yl)acetic acid
Synonyms: (7-amino-4-methyl-2-oxochromen-3-yl)acetic acid
Molecular Weight (g/mol): 233.22
Molecular Formula: C12H11NO4
InChi Key: QEQDLKUMPUDNPG-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)C(=O)OC2=CC(N)=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results