accessibility menu, dialog, popup

UserName

ONOHO

CAS RN 106276-04-4

IUPAC Name: {2-[2-(morpholin-4-yl)ethoxy]phenyl}methanol
Synonyms: {2-[2-(morpholin-4-yl)ethoxy]phenyl}methanol
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: MJDMCSJTOOAYPS-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1OCCN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)