accessibility menu, dialog, popup

UserName

CH3OONHNNNNNNNO

CAS RN 1062161-90-3

IUPAC Name: methyl N-{4-[4-(morpholin-4-yl)-1-{1-[(pyridin-3-yl)methyl]piperidin-4-yl}-1H-pyrazolo[3,4-d]pyrimidin-6-yl]phenyl}carbamate
Synonyms: methyl N-{4-[4-(morpholin-4-yl)-1-[1-(pyridin-3-ylmethyl)piperidin-4-yl]pyrazolo[3,4-d]pyrimidin-6-yl]phenyl}carbamate
Molecular Weight (g/mol): 528.62
Molecular Formula: C28H32N8O3
InChi Key: VDOCQQKGPJENHJ-UHFFFAOYSA-N
SMILES: COC(=O)NC1=CC=C(C=C1)C1=NC(N2CCOCC2)=C2C=NN(C3CCN(CC4=CN=CC=C4)CC3)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results