accessibility menu, dialog, popup

UserName

BrH2C

CAS RN 106-95-6

IUPAC Name: 3-bromoprop-1-ene
Synonyms: allyl bromide
Molecular Weight (g/mol): 120.98
Molecular Formula: C3H5Br
InChi Key: BHELZAPQIKSEDF-UHFFFAOYSA-N
SMILES: BrCC=C
Boiling Point 
  • (2)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (6)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)