accessibility menu, dialog, popup

UserName

CAS RN 106-89-8

IUPAC Name: 2-(chloromethyl)oxirane
Synonyms: epichlorohydrin caswell no. 424 (s)-epichlorohydrin skekhg α-epichlorohydrin (S)-isomer epichlorohydrin 3-chloro-1,2-propylene oxide epi-chlorohydrin epichloorhydrine epichlorohydrin (product)
Molecular Weight (g/mol): 92.52
Molecular Formula: C3H5ClO
InChi Key: BRLQWZUYTZBJKN-UHFFFAOYNA-N
SMILES: ClCC1CO1
Percent Purity 
  • (2)
  • (1)
  • (4)
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)