accessibility menu, dialog, popup

UserName

CAS RN 106-23-0

IUPAC Name: (3R)-3,7-dimethyloct-6-enal
Synonyms: (R)-(+)-citronellal (3R)-(+)-citronellal
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: NEHNMFOYXAPHSD-SNVBAGLBSA-N
SMILES: C[C@H](CCC=C(C)C)CC=O
Boiling Point 
  • (4)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)