accessibility menu, dialog, popup

UserName

CAS RN 105931-73-5

IUPAC Name: 4-bromo-2-fluoro-1-iodobenzene
Synonyms: 4-bromo-2-fluoro-1-iodobenzene
Molecular Weight (g/mol): 300.9
Molecular Formula: C6H3BrFI
InChi Key: XRMZKCQCINEBEI-UHFFFAOYSA-N
SMILES: FC1=C(I)C=CC(Br)=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)