accessibility menu, dialog, popup

UserName

CAS RN 10543-57-4

IUPAC Name: N-acetyl-N-[2-(N-acetylacetamido)ethyl]acetamide
Synonyms: TAED
Molecular Weight (g/mol): 228.25
Molecular Formula: C10H16N2O4
InChi Key: BGRWYDHXPHLNKA-UHFFFAOYSA-N
SMILES: CC(=O)N(CCN(C(C)=O)C(C)=O)C(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)