accessibility menu, dialog, popup

UserName

CAS RN 10519-33-2

H3CCH3O(E)

CAS RN 10519-33-2

IUPAC Name: (3E)-dec-3-en-2-one
Synonyms: (3E)-dec-3-en-2-one
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: JRPDANVNRUIUAB-CMDGGOBGSA-N
SMILES: CCCCCC\C=C\C(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results