accessibility menu, dialog, popup

UserName

H3COON

CAS RN 105-56-6

IUPAC Name: ethyl 2-cyanoacetate
Synonyms: ethyl cyanoacetate
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: ZIUSEGSNTOUIPT-UHFFFAOYSA-N
SMILES: CCOC(=O)CC#N
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)