accessibility menu, dialog, popup

UserName

CAS RN 105-45-3

IUPAC Name: methyl 3-oxobutanoate
Synonyms: methyl acetoacetate methylacetoacetate methyl acetylacetonate methylester kyseliny acetoctove
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: WRQNANDWMGAFTP-UHFFFAOYSA-N
SMILES: COC(=O)CC(C)=O
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
Boiling Point 
  • (4)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (7)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)