accessibility menu, dialog, popup

UserName

CAS RN 10493-44-4

IUPAC Name: 4-bromo-1,1,2-trifluorobut-1-ene
Synonyms: 4-bromo-1,1,2-trifluorobut-1-ene
Molecular Weight (g/mol): 188.98
Molecular Formula: C4H4BrF3
InChi Key: GQCQMFYIFUDARF-UHFFFAOYSA-N
SMILES: FC(F)=C(F)CCBr
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)