CAS RN 10476-85-4
IUPAC Name:
strontium(2+) hexahydrate dichloride
Synonyms:
strontium chloride
Molecular Weight (g/mol):
266.61
Molecular Formula:
Cl2H12O6Sr
InChi Key:
AMGRXJSJSONEEG-UHFFFAOYSA-L
SMILES:
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
1
–
5
of
5
results
1
Strontium chloride, anhydrous, 95%
CAS: 10476-85-4 MDL Number: MFCD00011249
2
Strontium chloride, 99.99%, (trace metal basis), anhydrous
CAS: 10476-85-4 Molecular Formula: Cl2Sr Molecular Weight (g/mol): 158.53 MDL Number: MFCD00011249
4