accessibility menu, dialog, popup

UserName

CAS RN 10439-77-7

H3CNHH3CNOO

CAS RN 10439-77-7

IUPAC Name: N,2-dimethyl-4-nitroaniline
Synonyms: N,2-dimethyl-4-nitroaniline
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: NJYYBTRBCMPYCJ-UHFFFAOYSA-N
SMILES: CNC1=CC=C(C=C1C)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results