CAS RN 104206-65-7
IUPAC Name:
2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione
Synonyms:
nitisinone nitisinonum nitisinona orfadin
Molecular Weight (g/mol):
329.23
Molecular Formula:
C14H10F3NO5
InChi Key:
OUBCNLGXQFSTLU-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(=CC=C1C(=O)C1C(=O)CCCC1=O)C(F)(F)F
1
–
2
of
2
results
1
Promotions Available
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 95%
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 95%
CAS: 104206-65-7 Molecular Formula: C14H10F3NO5 Molecular Weight (g/mol): 329.231 MDL Number: MFCD01752192 InChI Key: OUBCNLGXQFSTLU-UHFFFAOYSA-N Synonym: nitisinone,orfadin,nitisone,ntbc,2-2-nitro-4-trifluoromethyl benzoyl cyclohexane-1,3-dione,2-2-nitro-4-trifluoromethylbenzoyl-1,3-cyclohexanedione,nitisinone usan:inn,nitisinone inn,nitisinona PubChem CID: 115355 ChEBI: CHEBI:50378 IUPAC Name: 2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione SMILES: C1CC(=O)C(C(=O)C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-]