missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 95%

Catalog No. AAH66633MD
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66633MD 250 mg
AAH6663303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAH66633MD missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' H66633MD

CAS: 104206-65-7 | C14H10F3NO5 | 329.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104206-65-7
Molecular Formula C14H10F3NO5
Molecular Weight (g/mol) 329.231
MDL Number MFCD01752192
InChI Key OUBCNLGXQFSTLU-UHFFFAOYSA-N
Synonym nitisinone, orfadin, nitisone, ntbc, 2-2-nitro-4-trifluoromethyl benzoyl cyclohexane-1,3-dione, 2-2-nitro-4-trifluoromethylbenzoyl-1,3-cyclohexanedione, nitisinone usan:inn, nitisinone inn, nitisinona
PubChem CID 115355
ChEBI CHEBI:50378
IUPAC Name 2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione
SMILES C1CC(=O)C(C(=O)C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-]

Specifications

Quantity 250 mg
Formula Weight 329.23
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione
RTECSNumber GV0766666
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only