missing translation for 'onlineSavingsMsg'
Learn More

Nitisinone Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111012362
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012362 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012362 Supplier Merck Emd Millipore Supplier No. PHR17311G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Nitisinone is a medication, effectively used for the treatment and maintainence of alkaptonuria, an autosomal recessive disease. Its mode of action involves the inhibition of second enzyme in the tyrosine pathway, 4-hydroxyphenylpyruvate dioxygenase that produces HGA from 4-hydroxyphenylpyruvate.

Specifications

CAS 104206-65-7
Linear Formula C14H10F3NO5
MDL Number MFCD01752192
Synonym Nitisinone; 2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione; 2-[2-Nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione; NTBC; Nitisone; SC 0735
Quantity 1 g
Formula Weight 329.23
Grade Certified Reference Material