accessibility menu, dialog, popup

UserName

CH3OOH3CNN(E)

CAS RN 1041-00-5

IUPAC Name: 5-methyl-2-[(1E)-2-(5-methyl-1,3-benzoxazol-2-yl)ethenyl]-1,3-benzoxazole
Synonyms: 5-methyl-2-[(1E)-2-(5-methyl-1,3-benzoxazol-2-yl)ethenyl]-1,3-benzoxazole
Molecular Weight (g/mol): 290.32
Molecular Formula: C18H14N2O2
InChi Key: VKRZNAWSCAUDRQ-BQYQJAHWSA-N
SMILES: CC1=CC=C2OC(\C=C\C3=NC4=CC(C)=CC=C4O3)=NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results