accessibility menu, dialog, popup

UserName

CAS RN 104-84-7

IUPAC Name: 1-(4-methylphenyl)methanamine
Synonyms: benzenemethanamine, 4-methyl-
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: HMTSWYPNXFHGEP-UHFFFAOYSA-N
SMILES: CC1=CC=C(CN)C=C1
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)