missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzylamine, 98%

Catalog No. AAA1376822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1376822 100 g
AAA1376814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1376822 Supplier Thermo Scientific Chemicals Supplier No. A1376822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104-84-7 | C8H11N | 121.183 g/mol

The reactions of 1-hexanol with benzylamine and with 4-methylbenzylamine led to the corresponding imines in moderate yields. 4-methylbenzylamine in ethanol/water solution (1:4) as simultaneously absorbing and buffering background electrolyte with detection at 210 nm was found suitable for determination of the individual compounds in determination by capillary zone electrophoresis with indirect detection. Synthesis of benzimidazoles uses 4-methylbenzylamine (R 1 =4-MeBn) and 4-chlorophenyl isothiocyanate (R 2 =4-ClPh) as building blocks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The reactions of 1-hexanol with benzylamine and with 4-methylbenzylamine led to the corresponding imines in moderate yields. 4-methylbenzylamine in ethanol/water solution (1:4) as simultaneously absorbing and buffering background electrolyte with detection at 210 nm was found suitable for determination of the individual compounds in determination by capillary zone electrophoresis with indirect detection. Synthesis of benzimidazoles uses 4-methylbenzylamine (R 1 =4-MeBn) and 4-chlorophenyl isothiocyanate (R 2 =4-ClPh) as building blocks.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 104-84-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008123
InChI Key HMTSWYPNXFHGEP-UHFFFAOYSA-N
Synonym 4-methylbenzylamine, benzenemethanamine, 4-methyl, p-methylbenzylamine, p-tolylmethanamine, 4-methylphenyl methanamine, benzylamine, p-methyl, p-xylylamine, 4-methybenzylamine, 1-4-methylphenyl methanamine, 4-methyl-benzylamine
PubChem CID 66035
IUPAC Name (4-methylphenyl)methanamine
SMILES CC1=CC=C(C=C1)CN

Specifications

Melting Point 12°C to 13°C
Density 0.955
Boiling Point 196°C to 197°C
Flash Point 75°C (167°F)
Refractive Index 1.535
Quantity 100 g
UN Number UN2735
Beilstein 956670
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 121.18
Percent Purity 98%
Chemical Name or Material 4-Methylbenzylamine
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 203-243-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only