missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzylamine 98.0+%, TCI America™

Catalog No. M086425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M086425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M086425ML Supplier TCI America Supplier No. M086425ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104-84-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008123
InChI Key HMTSWYPNXFHGEP-UHFFFAOYSA-N
Synonym 4-methylbenzylamine, benzenemethanamine, 4-methyl, p-methylbenzylamine, p-tolylmethanamine, 4-methylphenyl methanamine, benzylamine, p-methyl, p-xylylamine, 4-methybenzylamine, 1-4-methylphenyl methanamine, 4-methyl-benzylamine
PubChem CID 66035
IUPAC Name (4-methylphenyl)methanamine
SMILES CC1=CC=C(C=C1)CN

Specifications

Melting Point 13°C
Color Yellow
Boiling Point 196°C
Quantity 25 mL
UN Number 2735
Formula Weight 121.18
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Methylbenzylamine