accessibility menu, dialog, popup

UserName

H3CCH3OH

CAS RN 104-76-7

IUPAC Name: 2-ethylhexan-1-ol
Synonyms: 2-ethylhexanol ethylhexanol 2-aethylhexanol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: YIWUKEYIRIRTPP-UHFFFAOYNA-N
SMILES: CCCCC(CC)CO
Molecular Weight (g/mol) 
  • (6)
  • (4)
Percent Purity 
  • (4)
  • (1)
  • (5)
Boiling Point 
  • (2)
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (2)
  • (5)
  • (6)
  • (4)
  • (4)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)