missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2-Ethyl-1-hexanol

Catalog No. 5380514L
Encompass
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5380514L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5380514L Supplier Sigma Aldrich Supplier No. 5380514L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Ethyl-1-hexanol

Specifications

CAS 104-76-7
Melting Point -76°C (lit.)
Density 0.833 g/mL (at 25°C (literature))
Boiling Point 183°C to 186°C (lit.)
Molecular Formula C8H18O
Refractive Index n20/D 1.431 (literature)
Linear Formula CH3(CH2)3 CH(C2H5)CH2OH
MDL Number MFCD00004746
Quantity 4 L
Synonym Isooctyl alcohol
Molecular Weight (g/mol) 130.23
Percent Purity ≥99.6%
Show More Show Less