missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1-hexanol, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020585
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020585 1 mL
111019155 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020585 Supplier Merck Emd Millipore Supplier No. 086071MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-Ethyl-1-hexanol is an industrial chemical widely used in the production of polyvinyl chloride(PVC) plasticizers and is a potent indoor air pollutant emitted mainly from furniture or carpets with a PVC backing.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Chemical Name or Material 2-Ethyl-1-hexanol
CAS 104-76-7
Ignition Point 550°F
Vapor Pressure 0.2 mmHg (20°C)
Vapor Density 4.49 (vs. air)
Density 0.833 g/mL (at 25°C)
Flash Point 75°C - closed cup
Linear Formula CH3(CH2)3 CH(C2H5)CH2 OH
Technique Gas Chromatography
MDL Number MFCD00004746
Melting Point -76°C
Boiling Point 183°C to 186°C
Synonym Isooctyl alcohol
Type Analytical Standard
Solvent or Matrix Neat
Industry Type Water Analysis
Molecular Weight (g/mol) 130.23
Quantity 1 mL
Formula Weight 130.23
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less